Product Details

SNP ID
rs1062986
Assay Type
Validated
NCBI dbSNP Submissions
43
Location
Chr.1:108136110 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCCTTCAGAAATTCCACCACTGTG[C/T]TCATTCGTATTCAGCTGAAAGAAAA
Phenotype
MIM: 608744
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A24 PubMed Links

Gene Details

Gene
SLC25A24
Gene Name
solute carrier family 25 member 24
There are no transcripts associated with this gene.

View Full Product Details