Product Details

SNP ID
rs2877339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45579520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCTCCCACTGGCCCGCTTATC[C/T]TCGTGGCCTGCAGCCTGGTCCTGTC
Phenotype
MIM: 103072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY1 PubMed Links

Gene Details

Gene
ADCY1
Gene Name
adenylate cyclase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281768.1 Intron NP_001268697.1
NM_021116.2 Intron NP_066939.1
XM_005249584.3 Intron XP_005249641.1
XM_005249585.2 Intron XP_005249642.1

View Full Product Details