Product Details

SNP ID
rs10793442
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:43556371 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAAGAGAAACCAGAGTTTCAATTT[A/C]TCAGAGAATGTGAACAAAGGGAAAC
Phenotype
MIM: 601069
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF239 PubMed Links

Gene Details

Gene
ZNF239
Gene Name
zinc finger protein 239
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099282.1 1880 UTR 3 NP_001092752.1
NM_001099283.1 1880 UTR 3 NP_001092753.1
NM_001099284.1 1880 UTR 3 NP_001092754.1
NM_001324347.1 1880 UTR 3 NP_001311276.1
NM_001324348.1 1880 UTR 3 NP_001311277.1
NM_001324349.1 1880 UTR 3 NP_001311278.1
NM_001324350.1 1880 UTR 3 NP_001311279.1
NM_001324351.1 1880 UTR 3 NP_001311280.1
NM_001324352.1 1880 UTR 3 NP_001311281.1
NM_001324353.1 1880 UTR 3 NP_001311282.1
NM_005674.2 1880 UTR 3 NP_005665.2
XM_005271832.2 1880 UTR 3 XP_005271889.1
XM_006718001.2 1880 UTR 3 XP_006718064.1
XM_006718003.3 1880 UTR 3 XP_006718066.1
XM_011540232.2 1880 UTR 3 XP_011538534.1
XM_011540234.2 1880 UTR 3 XP_011538536.1
XM_011540235.2 1880 UTR 3 XP_011538537.1
XM_011540236.2 1880 UTR 3 XP_011538538.1
XM_011540237.2 1880 UTR 3 XP_011538539.1
XM_011540238.2 1880 UTR 3 XP_011538540.1
XM_017016740.1 1880 UTR 3 XP_016872229.1

View Full Product Details