Product Details
- SNP ID
-
rs1143643
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112830725 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGTTGTAACTGGGCCCCCAACTTTC[C/T]GTGTGATTATTAAAAGAGCCCTTCT
- Phenotype
-
MIM: 147720
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL1B
PubMed Links
Gene Details
- Gene
- IL1B
- Gene Name
- interleukin 1 beta
View Full Product Details