Product Details

SNP ID
rs7290980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30696438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGGCCTCCCCAATGAATTTCTG[A/T]TACAGTGGGCCTAGGGGAAGGGCCT
Phenotype
MIM: 606729
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OSBP2 PubMed Links

Gene Details

Gene
OSBP2
Gene Name
oxysterol binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282738.1 Intron NP_001269667.1
NM_001282739.1 Intron NP_001269668.1
NM_001282740.1 Intron NP_001269669.1
NM_001282741.1 Intron NP_001269670.1
NM_001282742.1 Intron NP_001269671.1
NM_030758.3 Intron NP_110385.1
XM_005261465.4 Intron XP_005261522.1
XM_006724203.2 Intron XP_006724266.2
XM_006724207.2 Intron XP_006724270.1
XM_006724208.3 Intron XP_006724271.1
XM_011530057.1 Intron XP_011528359.1
XM_011530058.1 Intron XP_011528360.1
XM_011530059.2 Intron XP_011528361.1
XM_011530060.1 Intron XP_011528362.1
XM_011530061.1 Intron XP_011528363.1
XM_011530062.1 Intron XP_011528364.1
XM_017028708.1 Intron XP_016884197.1
XM_017028709.1 Intron XP_016884198.1
XM_017028710.1 Intron XP_016884199.1
XM_017028711.1 Intron XP_016884200.1
XM_017028712.1 Intron XP_016884201.1
XM_017028713.1 Intron XP_016884202.1

View Full Product Details