Product Details

SNP ID
rs28425623
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:206110952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAATAACCAAGCTCAGGCCCCG[T/C]CCATCTCCATCCCCACTTACATTAT
Phenotype
MIM: 600264 MIM: 616088
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
AVPR1B PubMed Links

Gene Details

Gene
AVPR1B
Gene Name
arginine vasopressin receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000707.3 Intron NP_000698.1
Gene
C1orf186
Gene Name
chromosome 1 open reading frame 186
There are no transcripts associated with this gene.

View Full Product Details