Product Details

SNP ID
rs4147727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206126260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTATTTGCCACTTTGGAAAATTC[A/G]CCATAAATTGCTACACAGTGACTTT
Phenotype
MIM: 157655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCSHP3 PubMed Links

Gene Details

Gene
GCSHP3
Gene Name
glycine cleavage system protein H pseudogene 3
There are no transcripts associated with this gene.

Gene
NDUFS1
Gene Name
NADH:ubiquinone oxidoreductase core subunit S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199981.1 Intron NP_001186910.1
NM_001199982.1 Intron NP_001186911.1
NM_001199983.1 Intron NP_001186912.1
NM_001199984.1 Intron NP_001186913.1
NM_005006.6 Intron NP_004997.4
XM_017004188.1 Intron XP_016859677.1

View Full Product Details