Product Details
- SNP ID
-
rs1570492
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:56461097 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAAAACACAGCCAGAGAACTATTTG[G/C]AGGAAATGCATTGAAACAAATTACA
- Phenotype
-
MIM: 113810
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DST
PubMed Links
Gene Details
- Gene
- DST
- Gene Name
- dystonin
View Full Product Details