Product Details

SNP ID
rs1462919
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:78870512 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTTGAAATCAAAGATAAAATGCAC[A/T]TATGCACTCAAATGCAAACATATTT
Phenotype
MIM: 185605
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SYT1 PubMed Links

Gene Details

Gene
SYT1
Gene Name
synaptotagmin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135805.1 Intron NP_001129277.1
NM_001135806.1 Intron NP_001129278.1
NM_001291901.1 Intron NP_001278830.1
NM_005639.2 Intron NP_005630.1
XM_005269113.2 Intron XP_005269170.1
XM_006719576.1 Intron XP_006719639.1
XM_011538710.1 Intron XP_011537012.1
XM_017019909.1 Intron XP_016875398.1

View Full Product Details