Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032773.3 | Intron | NP_116162.1 | |||
XM_005269362.2 | Intron | XP_005269419.1 | |||
XM_005269363.2 | Intron | XP_005269420.1 | |||
XM_005269365.2 | Intron | XP_005269422.1 | |||
XM_005269367.2 | Intron | XP_005269424.1 | |||
XM_006713791.3 | Intron | XP_006713854.1 | |||
XM_006713792.3 | Intron | XP_006713855.1 | |||
XM_006713793.2 | Intron | XP_006713856.1 | |||
XM_011513242.2 | Intron | XP_011511544.1 | |||
XM_011513243.2 | Intron | XP_011511545.1 | |||
XM_017007350.1 | Intron | XP_016862839.1 | |||
XM_017007351.1 | Intron | XP_016862840.1 | |||
XM_017007352.1 | Intron | XP_016862841.1 |