Product Details
- SNP ID
-
rs11799942
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
43
- Location
-
Chr.1:145749827 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCTTAAATAGAATGACCTAAGAT[G/A]GCCCACCTTGTCTTTAAAATCTCTA
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF115
PubMed Links
Gene Details
- Gene
- RNF115
- Gene Name
- ring finger protein 115
View Full Product Details