Product Details
- SNP ID
-
rs2306421
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:121670198 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCAAGTATTTTCTTAGACACTGGCT[C/T]AGGTTTTCCTTGGAGTTCTGAGGTC
- Phenotype
-
MIM: 131230
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA5
PubMed Links
Gene Details
- Gene
- ANXA5
- Gene Name
- annexin A5
View Full Product Details