Product Details
- SNP ID
-
rs441687
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55844744 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTTGTGACCTCCAGTGTAACTCA[C/T]TGTATCTACGAGTATGCTATTTTCA
- Phenotype
-
MIM: 609664
MIM: 609645
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NLRP11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114009946] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NLRP11
- Gene Name
- NLR family pyrin domain containing 11
There are no transcripts associated with this gene.
- Gene
- NLRP4
- Gene Name
- NLR family pyrin domain containing 4
View Full Product Details