Product Details

SNP ID
rs1284532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53508635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATCAGGATGAAATCACTGGGGG[C/T]TATGAATCTATTTCCAAAATATGAG
Phenotype
MIM: 606043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF331 PubMed Links

Gene Details

Gene
ZNF331
Gene Name
zinc finger protein 331
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079906.1 Intron NP_001073375.1
NM_001079907.1 Intron NP_001073376.1
NM_001253798.1 Intron NP_001240727.1
NM_001253799.1 Intron NP_001240728.1
NM_001253800.1 Intron NP_001240729.1
NM_001253801.2 Intron NP_001240730.1
NM_001317113.1 Intron NP_001304042.1
NM_001317114.1 Intron NP_001304043.1
NM_001317115.1 Intron NP_001304044.1
NM_001317116.1 Intron NP_001304045.1
NM_001317117.1 Intron NP_001304046.1
NM_001317118.1 Intron NP_001304047.1
NM_001317119.1 Intron NP_001304048.1
NM_001317120.1 Intron NP_001304049.1
NM_001317121.1 Intron NP_001304050.1
NM_018555.5 Intron NP_061025.5
XM_011527076.2 Intron XP_011525378.1
XM_011527078.2 Intron XP_011525380.1
XM_017026936.1 Intron XP_016882425.1
XM_017026937.1 Intron XP_016882426.1
XM_017026938.1 Intron XP_016882427.1
XM_017026939.1 Intron XP_016882428.1
XM_017026940.1 Intron XP_016882429.1

View Full Product Details