Product Details

SNP ID
rs4985957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21002944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGGCCGGGGAGGCGGCGGGAG[G/A]CTTGGGGGAGGGGGGGGCCAGGGAG
Phenotype
MIM: 612116
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC105369205 PubMed Links

Gene Details

Gene
LOC105369205
Gene Name
uncharacterized LOC105369205
There are no transcripts associated with this gene.

Gene
USP22
Gene Name
ubiquitin specific peptidase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015276.1 1869 UTR 3 NP_056091.1
XM_005256575.2 1869 UTR 3 XP_005256632.1

View Full Product Details