Product Details
- SNP ID
-
rs9565177
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:75621267 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGATGGAATGAATAGATTGCATT[G/T]TGTTTCCATGCACAGGGGAACTGTG
- Phenotype
-
MIM: 604362
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LMO7
PubMed Links
Gene Details
- Gene
- LMO7
- Gene Name
- LIM domain 7
- Gene
- LMO7-AS1
- Gene Name
- LMO7 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details