Product Details

SNP ID
rs9544008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:75623761 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAATGAAAACTTTGGCTGATGAA[A/G]TTTACCAATTTAGATTTGTTTTCAG
Phenotype
MIM: 604362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMO7 PubMed Links

Gene Details

Gene
LMO7
Gene Name
LIM domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306080.1 Intron NP_001293009.1
NM_005358.5 Intron NP_005349.3
NM_015842.2 Intron NP_056667.2
XM_011535075.2 Intron XP_011533377.1
XM_011535078.1 Intron XP_011533380.1
XM_011535080.2 Intron XP_011533382.1
XM_011535082.1 Intron XP_011533384.1
XM_011535086.1 Intron XP_011533388.1
XM_011535094.1 Intron XP_011533396.1
XM_011535095.1 Intron XP_011533397.1
XM_017020574.1 Intron XP_016876063.1
XM_017020575.1 Intron XP_016876064.1
XM_017020576.1 Intron XP_016876065.1
XM_017020577.1 Intron XP_016876066.1
XM_017020578.1 Intron XP_016876067.1
XM_017020579.1 Intron XP_016876068.1
XM_017020580.1 Intron XP_016876069.1
XM_017020581.1 Intron XP_016876070.1
XM_017020582.1 Intron XP_016876071.1
XM_017020583.1 Intron XP_016876072.1
XM_017020584.1 Intron XP_016876073.1
XM_017020585.1 Intron XP_016876074.1
XM_017020586.1 Intron XP_016876075.1
XM_017020587.1 Intron XP_016876076.1
XM_017020588.1 Intron XP_016876077.1
XM_017020589.1 Intron XP_016876078.1
XM_017020590.1 Intron XP_016876079.1
XM_017020591.1 Intron XP_016876080.1
XM_017020592.1 Intron XP_016876081.1
XM_017020593.1 Intron XP_016876082.1
XM_017020594.1 Intron XP_016876083.1
XM_017020595.1 Intron XP_016876084.1
XM_017020596.1 Intron XP_016876085.1
XM_017020597.1 Intron XP_016876086.1
XM_017020598.1 Intron XP_016876087.1
XM_017020599.1 Intron XP_016876088.1
XM_017020600.1 Intron XP_016876089.1
XM_017020601.1 Intron XP_016876090.1
XM_017020602.1 Intron XP_016876091.1
XM_017020603.1 Intron XP_016876092.1
XM_017020604.1 Intron XP_016876093.1
Gene
LMO7-AS1
Gene Name
LMO7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details