Product Details

SNP ID
rs34224681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:98451139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGTGCTAAAAACTGTGATGTC[A/G]TTGTCCATTTGTAAATCTGAAGAAC
Phenotype
MIM: 602654 MIM: 604984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FARP1 PubMed Links

Gene Details

Gene
FARP1
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 1
There are no transcripts associated with this gene.

Gene
STK24
Gene Name
serine/threonine kinase 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032296.3 3695 UTR 3 NP_001027467.2
NM_001286649.1 3695 UTR 3 NP_001273578.1
NM_003576.4 3695 UTR 3 NP_003567.2
XM_005254079.4 3695 Intron XP_005254136.1
XM_017020794.1 3695 Intron XP_016876283.1
XM_017020795.1 3695 Intron XP_016876284.1

View Full Product Details