Product Details
- SNP ID
-
rs10146870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55579214 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACGAAACTCACTTGCCAGTGGAC[C/G]GGGAGGATCGCTGCCATCCTTTCAG
- Phenotype
-
MIM: 600381
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KTN1
PubMed Links
Gene Details
- Gene
- KTN1
- Gene Name
- kinectin 1
- Gene
- KTN1-AS1
- Gene Name
- KTN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details