Product Details

SNP ID
rs7983986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:37003588 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCTGTGCTTCTTTGACTACAGT[A/C]TACCCTGTGTTATTTAACCTCAGTT
Phenotype
MIM: 604565 MIM: 606019 MIM: 613417
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALG5 PubMed Links

Gene Details

Gene
ALG5
Gene Name
ALG5, dolichyl-phosphate beta-glucosyltransferase
There are no transcripts associated with this gene.

Gene
EXOSC8
Gene Name
exosome component 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181503.2 Intron NP_852480.1
XM_006719763.1 Intron XP_006719826.1
Gene
SUPT20H
Gene Name
SPT20 homolog, SAGA complex component
There are no transcripts associated with this gene.

View Full Product Details