Product Details

SNP ID
rs10491742
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:12775489 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGAGATCTTGATCATCTTAGTGTC[A/G]GAGGAGTCGCAGCGGACTGGGAACT
Phenotype
MIM: 616130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LURAP1L PubMed Links

Gene Details

Gene
LURAP1L
Gene Name
leucine rich adaptor protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203403.1 477 UTR 5 NP_981948.1
XM_005251443.2 477 UTR 5 XP_005251500.1
Gene
LURAP1L-AS1
Gene Name
LURAP1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details