Product Details

SNP ID
rs35921123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13282670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACATGATTTGTGTCGTTGTGC[C/T]TGCACCTCGCTACTTGCACAGCTCT
Phenotype
MIM: 602026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHYH PubMed Links

Gene Details

Gene
PHYH
Gene Name
phytanoyl-CoA 2-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037537.1 Intron NP_001032626.1
NM_001323080.1 Intron NP_001310009.1
NM_001323082.1 Intron NP_001310011.1
NM_001323083.1 Intron NP_001310012.1
NM_001323084.1 Intron NP_001310013.1
NM_006214.3 Intron NP_006205.1

View Full Product Details