Product Details

SNP ID
rs11617415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20142272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACGCTGCCGGGTAAGCCTTGAG[C/T]GCCGGGGGCTGCCGCTCGGCCGCCT
Phenotype
MIM: 121015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA3 PubMed Links

Gene Details

Gene
GJA3
Gene Name
gap junction protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021954.3 1096 Silent Mutation GCC,GCT A339A NP_068773.2
XM_011535048.2 1096 Silent Mutation GCC,GCT A339A XP_011533350.1

View Full Product Details