Product Details

SNP ID
rs1048155
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:26120155 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACATCAAGCCAATGACAAAAAAAAG[C/G]TGTAAATCTGAGAACTGTAGTGATT
Phenotype
MIM: 606200 MIM: 601599
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BHLHE41 PubMed Links

Gene Details

Gene
BHLHE41
Gene Name
basic helix-loop-helix family member e41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030762.2 3651 UTR 3 NP_110389.1
Gene
SSPN
Gene Name
sarcospan
There are no transcripts associated with this gene.

View Full Product Details