Product Details

SNP ID
rs2721197
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:144502322 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTGGGCCTCACCCACTCCCTCCC[A/G]TGCTCAGCCTCCTGGAGCCTCCTCC
Phenotype
MIM: 138200 MIM: 609172 MIM: 603780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPT PubMed Links

Gene Details

Gene
GPT
Gene Name
glutamic--pyruvic transaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005309.2 106 Intron NP_005300.1
XM_011516993.2 106 UTR 5 XP_011515295.1
Gene
LOC101928953
Gene Name
uncharacterized LOC101928953
There are no transcripts associated with this gene.

Gene
MFSD3
Gene Name
major facilitator superfamily domain containing 3
There are no transcripts associated with this gene.

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272350.2 106 Intron XP_005272407.1
XM_011517349.2 106 Intron XP_011515651.1
XM_017013912.1 106 Intron XP_016869401.1
XM_017013913.1 106 Intron XP_016869402.1
XM_017013914.1 106 Intron XP_016869403.1
XM_017013915.1 106 Intron XP_016869404.1
Gene
RECQL4
Gene Name
RecQ like helicase 4
There are no transcripts associated with this gene.

View Full Product Details