Product Details

SNP ID
rs7330874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95677821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGCTGGGAGTTGGAGGGGACAG[C/T]GTTGACTATTTAAAAAGTGTGTAAC
Phenotype
MIM: 601184
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC3 PubMed Links

Gene Details

Gene
DNAJC3
Gene Name
DnaJ heat shock protein family (Hsp40) member C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006260.4 Intron NP_006251.1
XM_011521104.2 Intron XP_011519406.1
XM_011521105.1 Intron XP_011519407.1
XM_017020674.1 Intron XP_016876163.1
XM_017020675.1 Intron XP_016876164.1
Gene
DNAJC3-AS1
Gene Name
DNAJC3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details