Product Details
- SNP ID
-
rs617698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
63
- Location
-
Chr.1:160193082 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGGAGACTCTGCCTCTCTATTCCTT[A/G]TGAAGTTCCATGCTCAAATCCCAGA
- Phenotype
-
MIM: 607321
MIM: 114250
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP1A4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74337382] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP1A4
- Gene Name
- ATPase Na+/K+ transporting subunit alpha 4
There are no transcripts associated with this gene.
- Gene
- CASQ1
- Gene Name
- calsequestrin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001231.4 |
|
Intron |
|
|
NP_001222.3 |
- Gene
- LOC729867
- Gene Name
- uncharacterized LOC729867
There are no transcripts associated with this gene.
View Full Product Details