Product Details
- SNP ID
-
rs1061511
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
72
- Location
-
Chr.1:160219710 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAACAAGGCAAAAGAAATAAACT[A/C]CAGCAGGCAGGCCTTCTAGCTTAGG
- Phenotype
-
MIM: 615820
MIM: 603434
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DCAF8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145648849] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCAF8
- Gene Name
- DDB1 and CUL4 associated factor 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015726.3 |
|
Intron |
|
|
NP_056541.2 |
- Gene
- PEA15
- Gene Name
- phosphoprotein enriched in astrocytes 15
There are no transcripts associated with this gene.
View Full Product Details