Product Details

SNP ID
rs7290
Assay Type
Validated
NCBI dbSNP Submissions
64
Location
Chr.1:1541864 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AAGAAAAACTTTGTAATCAATATCC[T/C]GCTCATAAGTAAAAGTGGAAAAGAA
Phenotype
MIM: 612316 MIM: 616101
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ATAD3A PubMed Links

Gene Details

Gene
ATAD3A
Gene Name
ATPase family, AAA domain containing 3A
There are no transcripts associated with this gene.

Gene
SSU72
Gene Name
SSU72 homolog, RNA polymerase II CTD phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014188.2 1112 UTR 3 NP_054907.1
Gene
TMEM240
Gene Name
transmembrane protein 240
There are no transcripts associated with this gene.

View Full Product Details