Product Details

SNP ID
rs3774634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45944950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATACAGTGCCCCAGAGAGCTTGT[C/G]GGGGGCAGGATGTCTACTGGCTGAC
Phenotype
MIM: 605163 MIM: 607182
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXCR6 PubMed Links

Gene Details

Gene
CXCR6
Gene Name
C-X-C motif chemokine receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006564.1 Intron NP_006555.1
XM_005264809.2 Intron XP_005264866.1
XM_011533290.2 Intron XP_011531592.1
XM_011533291.2 Intron XP_011531593.1
Gene
FYCO1
Gene Name
FYVE and coiled-coil domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024513.3 Intron NP_078789.2
XM_006713333.3 Intron XP_006713396.1
XM_006713334.3 Intron XP_006713397.1
XM_011534111.2 Intron XP_011532413.1

View Full Product Details