Product Details

SNP ID
rs2634681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48339301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTGAGGTACCACTGCTTGAAAT[G/A]TGAATCCCCTAGCCAGTGGTTCTCA
Phenotype
MIM: 613906
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
H1FNT PubMed Links

Gene Details

Gene
H1FNT
Gene Name
H1 histone family member N, testis specific
There are no transcripts associated with this gene.

Gene
ZNF641
Gene Name
zinc finger protein 641
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172681.1 4922 Intron NP_001166152.1
NM_001172682.1 4922 Intron NP_001166153.1
NM_152320.2 4922 Intron NP_689533.2
XM_005268637.3 4922 UTR 3 XP_005268694.1
XM_005268638.3 4922 UTR 3 XP_005268695.1
XM_005268639.2 4922 UTR 3 XP_005268696.1
XM_005268640.4 4922 Intron XP_005268697.1
XM_011537897.1 4922 UTR 3 XP_011536199.1
XM_011537898.1 4922 UTR 3 XP_011536200.1
XM_017018797.1 4922 UTR 3 XP_016874286.1
XM_017018798.1 4922 UTR 3 XP_016874287.1
XM_017018799.1 4922 Intron XP_016874288.1

View Full Product Details