Product Details
- SNP ID
-
rs8103698
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56089929 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCCCAGCTCATTCACTCCTGAGAAG[A/G]GTCCGTGTGCAGCTTCTACATTTGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF787
PubMed Links
Gene Details
- Gene
- ZNF787
- Gene Name
- zinc finger protein 787
View Full Product Details