Product Details

SNP ID
rs12292156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76872005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTGAACTCCAAAGAGTGTAATGA[A/G]GCATATCTGACCCCTCTCCCCACCC
Phenotype
MIM: 617036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACER3 PubMed Links

Gene Details

Gene
ACER3
Gene Name
alkaline ceramidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300953.1 Intron NP_001287882.1
NM_001300954.1 Intron NP_001287883.1
NM_001300955.1 Intron NP_001287884.1
NM_018367.6 Intron NP_060837.3
XM_011545151.2 Intron XP_011543453.1
XM_011545152.2 Intron XP_011543454.1
XM_011545153.2 Intron XP_011543455.1
XM_017017987.1 Intron XP_016873476.1
XM_017017988.1 Intron XP_016873477.1

View Full Product Details