Product Details

SNP ID
rs6572596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49647114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATCTATTTCAACACAAACGTACC[A/G]TCATGTACCCACAATTCTGTGTCAC
Phenotype
MIM: 602670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLE2 PubMed Links

Gene Details

Gene
POLE2
Gene Name
DNA polymerase epsilon 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197330.1 Intron NP_001184259.1
NM_001197331.1 Intron NP_001184260.1
NM_002692.3 Intron NP_002683.2
XM_011536842.2 Intron XP_011535144.1
XM_011536843.2 Intron XP_011535145.1
XM_011536844.1 Intron XP_011535146.1
XM_017021374.1 Intron XP_016876863.1
XM_017021375.1 Intron XP_016876864.1

View Full Product Details