Product Details

SNP ID
rs4517109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74242708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTCGCGTGAAGGTGTCAAGTCG[A/G]TCTTGAATTTTGCAGAGGCAGTCTA
Phenotype
MIM: 605266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JPH1 PubMed Links

Gene Details

Gene
JPH1
Gene Name
junctophilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317830.1 Intron NP_001304759.1
NM_020647.3 Intron NP_065698.1
XM_005251274.3 Intron XP_005251331.1
XM_005251275.4 Intron XP_005251332.1

View Full Product Details