Product Details

SNP ID
rs329390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138063699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGGCAGCAAGCATGCTCTGAGC[A/G]GCGCCTCAATCTGTTTGGGGCTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DZIP1L PubMed Links

Gene Details

Gene
DZIP1L
Gene Name
DAZ interacting zinc finger protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170538.1 Intron NP_001164009.1
NM_173543.2 Intron NP_775814.2
XM_005247198.3 Intron XP_005247255.1
XM_005247200.4 Intron XP_005247257.1
XM_006713526.1 Intron XP_006713589.1
XM_006713527.3 Intron XP_006713590.1
XM_011512518.2 Intron XP_011510820.1
XM_011512520.2 Intron XP_011510822.1
XM_011512521.2 Intron XP_011510823.1
XM_011512522.1 Intron XP_011510824.1
XM_011512524.2 Intron XP_011510826.1
XM_011512528.2 Intron XP_011510830.1
XM_017005841.1 Intron XP_016861330.1
XM_017005842.1 Intron XP_016861331.1

View Full Product Details