Product Details
- SNP ID
-
rs1351902
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:152538373 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTCCAAAAAATCTGAAGTTTTAGCT[A/T]ATAGTTACTTCCCTGGTTCATCAAT
- Phenotype
-
MIM: 606278
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DKFZP434I0714
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75092985] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DKFZP434I0714
- Gene Name
- uncharacterized protein DKFZP434I0714
There are no transcripts associated with this gene.
- Gene
- FBXW7
- Gene Name
- F-box and WD repeat domain containing 7
There are no transcripts associated with this gene.
- Gene
- MIR4453
- Gene Name
- microRNA 4453
There are no transcripts associated with this gene.
View Full Product Details