Product Details

SNP ID
rs2057592
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:116997764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGATGTAAGTAGAAGCTTTAAAAA[A/T]TGCTGTTGTGTTTCCACTAGTTCTT
Phenotype
MIM: 604516 MIM: 601204
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD101 PubMed Links

Gene Details

Gene
CD101
Gene Name
CD101 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256106.2 Intron NP_001243035.1
NM_001256109.2 Intron NP_001243038.1
NM_001256111.2 Intron NP_001243040.1
NM_004258.5 Intron NP_004249.2
XM_017002846.1 Intron XP_016858335.1
XM_017002847.1 Intron XP_016858336.1
Gene
PTGFRN
Gene Name
prostaglandin F2 receptor inhibitor
There are no transcripts associated with this gene.

View Full Product Details