Product Details
- SNP ID
-
rs822875
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:29502011 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATATTTTCCTTTTGCATCAAAAAC[G/T]TCAAGAATAATCATAAAATCTTATA
- Phenotype
-
MIM: 615855
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OVCH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138577490] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OVCH1
- Gene Name
- ovochymase 1
There are no transcripts associated with this gene.
- Gene
- TMTC1
- Gene Name
- transmembrane and tetratricopeptide repeat containing 1
View Full Product Details