Product Details
- SNP ID
-
rs3750501
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:12775890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGGCGGCGGCGGCTGCAGTAGC[A/G]GCAGCAGCTACTGCAGCTTCCCTCC
- Phenotype
-
MIM: 616130
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LURAP1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75227666] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LURAP1L
- Gene Name
- leucine rich adaptor protein 1 like
- Gene
- LURAP1L-AS1
- Gene Name
- LURAP1L antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details