Product Details

SNP ID
rs6427641
Assay Type
Validated
NCBI dbSNP Submissions
55
Location
Chr.1:154408010 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCTGCCCCCATTACCCACACAC[G/A]TGCCTAGCCTCACCCTAAATCAGTA
Phenotype
MIM: 147880
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IL6R PubMed Links

Gene Details

Gene
IL6R
Gene Name
interleukin 6 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000565.3 Intron NP_000556.1
NM_001206866.1 Intron NP_001193795.1
NM_181359.2 Intron NP_852004.1
XM_005245139.1 Intron XP_005245196.1
XM_005245140.2 Intron XP_005245197.1
XM_006711298.1 Intron XP_006711361.1
XM_006711299.3 Intron XP_006711362.1
XM_017001199.1 Intron XP_016856688.1
XM_017001200.1 Intron XP_016856689.1
XM_017001201.1 Intron XP_016856690.1
Gene
LOC101928101
Gene Name
uncharacterized LOC101928101
There are no transcripts associated with this gene.

View Full Product Details