Product Details
- SNP ID
-
rs7342066
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:7753948 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAGAGAGATCCCAGGGTTTGGCACC[A/G]TACCTGTGTCTGACGTCAGCTTATA
- Phenotype
-
MIM: 146640
MIM: 601720
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITIH2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7067698] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITIH2
- Gene Name
- inter-alpha-trypsin inhibitor heavy chain 2
There are no transcripts associated with this gene.
- Gene
- KIN
- Gene Name
- Kin17 DNA and RNA binding protein
View Full Product Details