Product Details

SNP ID
rs10814574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37512987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTCTGTAGGCCTCAATTTTCCC[A/G]CTTTGAGACACTCTGTTGCCCAGGC
Phenotype
MIM: 609092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO10 PubMed Links

Gene Details

Gene
FBXO10
Gene Name
F-box protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012166.2 Intron NP_036298.2
XM_005251439.4 Intron XP_005251496.1
XM_006716754.3 Intron XP_006716817.1
XM_017014618.1 Intron XP_016870107.1
XM_017014619.1 Intron XP_016870108.1
Gene
POLR1E
Gene Name
RNA polymerase I subunit E
There are no transcripts associated with this gene.

View Full Product Details