Product Details

SNP ID
rs2800953
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:151366094 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTAAGTAGTTTTTGAATGAATG[A/G]ATAAATACGGCCAGTTAGGGTTCAG
Phenotype
MIM: 604188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 Intron NP_001245217.1
NM_001258289.1 Intron NP_001245218.1
NM_003944.3 Intron NP_003935.2
XM_017002757.1 Intron XP_016858246.1

View Full Product Details