Product Details

SNP ID
rs7138000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52076325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGATGCAATGCAAAGGTGAAAAG[C/G]AAGGATTTAGAAATCAGACAATCCA
Phenotype
MIM: 615089
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATG101 PubMed Links

Gene Details

Gene
ATG101
Gene Name
autophagy related 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098673.1 Intron NP_001092143.1
NM_021934.4 Intron NP_068753.2
Gene
OR7E47P
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
There are no transcripts associated with this gene.

View Full Product Details