Product Details

SNP ID
rs1053468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160563230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACTAAGTACAATCATTAGCATTA[C/T]GTTATAGGGGAATAGTGGTTATAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP10B PubMed Links

Gene Details

Gene
ATP10B
Gene Name
ATPase phospholipid transporting 10B (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025153.2 7013 UTR 3 NP_079429.2
XM_006714833.2 7013 UTR 3 XP_006714896.1
XM_011534468.2 7013 UTR 3 XP_011532770.1
XM_011534469.1 7013 UTR 3 XP_011532771.1
XM_011534472.2 7013 UTR 3 XP_011532774.1
XM_017009252.1 7013 UTR 3 XP_016864741.1
XM_017009253.1 7013 UTR 3 XP_016864742.1

View Full Product Details