Product Details

SNP ID
rs2227270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18309894 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGGGAGCAGGTTATTAACTTAC[A/C]ACCGGGCCGTCTGGCCCAGCCCTGG
Phenotype
MIM: 607284
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LSM4 PubMed Links

Gene Details

Gene
LSM4
Gene Name
LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252129.1 Intron NP_001239058.1
NM_012321.4 Intron NP_036453.1

View Full Product Details