Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001104586.2 | Intron | NP_001098056.1 | |||
NM_001282910.1 | Intron | NP_001269839.1 | |||
NM_001282911.1 | Intron | NP_001269840.1 | |||
NM_001282912.1 | Intron | NP_001269841.1 | |||
NM_001322427.1 | Intron | NP_001309356.1 | |||
NM_001322428.1 | Intron | NP_001309357.1 | |||
NM_001322429.1 | Intron | NP_001309358.1 | |||
NM_001322430.1 | Intron | NP_001309359.1 | |||
NM_024744.15 | Intron | NP_079020.13 | |||
XM_005246858.3 | Intron | XP_005246915.1 | |||
XM_005246859.3 | Intron | XP_005246916.1 | |||
XM_005246864.3 | Intron | XP_005246921.1 | |||
XM_006712760.2 | Intron | XP_006712823.1 | |||
XM_011511867.2 | Intron | XP_011510169.1 | |||
XM_011511868.2 | Intron | XP_011510170.1 | |||
XM_011511869.2 | Intron | XP_011510171.1 | |||
XM_017004961.1 | Intron | XP_016860450.1 | |||
XM_017004962.1 | Intron | XP_016860451.1 | |||
XM_017004963.1 | Intron | XP_016860452.1 | |||
XM_017004964.1 | Intron | XP_016860453.1 | |||
XM_017004965.1 | Intron | XP_016860454.1 | |||
XM_017004966.1 | Intron | XP_016860455.1 |