Product Details
- SNP ID
-
rs891394
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:27462252 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGAACTTGGGCTCACAGCCCAGA[C/G]AAGGGAGCCTGGATTCTCCACTGCT
- Phenotype
-
MIM: 118502
MIM: 601212
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CHRNA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77229364] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHRNA2
- Gene Name
- cholinergic receptor nicotinic alpha 2 subunit
- Gene
- PTK2B
- Gene Name
- protein tyrosine kinase 2 beta
There are no transcripts associated with this gene.
View Full Product Details