Product Details

SNP ID
rs11825599
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19184478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCGACAACAGAACTGTTAGTG[C/T]GGGGTAGGCAGGAGGGAGATAGACA
Phenotype
MIM: 600824 MIM: 612815
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CSRP3 PubMed Links

Gene Details

Gene
CSRP3
Gene Name
cysteine and glycine rich protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003476.4 Intron NP_003467.1
Gene
ZDHHC13
Gene Name
zinc finger DHHC-type containing 13
There are no transcripts associated with this gene.

View Full Product Details